2-Acetyl-4-bromothiophene
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10X-0139.1
10X-0139.10
10X-0139.5
10X-0139.500
BIOT10X-0139.1EA
0
GBP
BIOT10X-0139.1
BIOT10X-0139.10
BIOT10X-0139.5
BIOT10X-0139.500
2-Acetyl-4-bromothiophene
2-Acetyl-4-bromothiophene
Formula:
C₆H₅BrOS MW: 205.08 g/mol |
MDL Number:
MFCD01566552 CAS Number: 7209-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...