2-Chloroisonicotinonitrile ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Chloro-4-cyanopyridine, 2-Chloropyridine-4-carbonitrile

L19858.06 L19858.14
L19858.06EA 29.5 GBP
L19858.06 L19858.14
2-Chloroisonicotinonitrile ≥97%
2-Chloroisonicotinonitrile

Purity: 97% CAS No.: 33252-30-1 Molecular Formula: C6H3ClN2 Formula Weight: 138.56 Melting Point: 69-72° Boiling Point: 104-106°/15mm UN No.: 3439 MDL No.: MFCD00174318

Formula: C₆H₃ClN₂
MW: 138.56 g/mol
Boiling Pt: 104…106 °C (15 mmHg)
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00174318
CAS Number: 33252-30-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR