2-Acetyl-5-iodothiophene ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(5-Iodo-2-thienyl)-1-ethanone, 1-(5-Iodothiophen-2-yl)ethanone

L17852.03 L17852.06
L17852.03EA 0 GBP
L17852.03 L17852.06
2-Acetyl-5-iodothiophene ≥97%
2-Acetyl-5-iodothiophene

Purity: 97% CAS No.: 30955-94-3 Molecular Formula: C6H5IOS Formula Weight: 252.07 Melting Point: 127-129° MDL No.: MFCD02093967

Formula: C₆H₅IOS
MW: 252.08 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD02093967
CAS Number: 30955-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR