2-Amino-5-fluorophenol ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H32928.MD
H32928.03
H32928.MDEA
0
GBP
H32928.MD
H32928.03
2-Amino-5-fluorophenol ≥97%
2-Amino-5-fluorophenol
Purity: 97%
CAS No.: 53981-24-1
Molecular Formula: C6H6FNO
Formula Weight: 127.12
Storage Temperature: 2°C to 8°C
Melting Point: 135-140°
MDL No.: MFCD00671759
Formula:
C₆H₆FNO MW: 127.12 g/mol Melting Pt: 135…140 °C |
MDL Number:
MFCD00671759 CAS Number: 53981-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...