2-Chloropyridine-6-carbonitrile ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
6-Chloropicolinonitrile, 6-Chloropyridine-2-carbonitrile, 2-Chloro-6-cyanopyridine
H27748.03
H27748.06
H27748.03EA
32.5
GBP
H27748.03
H27748.06
2-Chloropyridine-6-carbonitrile ≥97%
2-Chloropyridine-6-carbonitrile
Purity: 97%
CAS No.: 33252-29-8
Molecular Formula: C6H3ClN2
Formula Weight: 138.56
Melting Point: 85-88°
UN No.: 3439
MDL No.: MFCD00274527
Formula:
C₆H₃ClN₂ MW: 138.56 g/mol Melting Pt: 85…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274527 CAS Number: 33252-29-8 EINECS: 251-429-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...