Potassium tetrabromoplatinate(II) ≥99%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B23308.88EA 0 GBP
B23308.88
Potassium tetrabromoplatinate(II) ≥99%
Potassium tetrabromoplatinate (II)
Formula: Br₄K₂Pt
MW: 592.89 g/mol
MDL Number: MFCD00011376
CAS Number: 13826-94-3
EINECS: 237-536-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR