5-Amino-2-methylindole 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2-Methyl-5-indolamine, 2-Methylindol-5-ylamine, 5-Amino-2-methyl-1H-indole
B21694.03
B21694.06
B21694.14
B21694.03EA
29.7
GBP
B21694.03
B21694.06
B21694.14
5-Amino-2-methylindole 97%
5-Amino-2-methylindole
Purity: 97%
CAS No.: 7570-49-2
Molecular Formula: C9H10N2
Formula Weight: 145.19
Storage Temperature: 2°C to 8°C
Formula:
C₉H₁₀N₂ MW: 146.19 g/mol Melting Pt: 158…159 °C |
MDL Number:
MFCD02093426 CAS Number: 7570-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...