Trimethylammonium chloride 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Trimethylamine hydrochloride

ACRO140071000EA 20.1 GBP
ACRO140071000
Trimethylammonium chloride 98%
Trimethylammonium chloride
Formula: C₃H₁₀ClN
MW: 95.57 g/mol
Melting Pt: 283…284 °C
MDL Number: MFCD00012478
CAS Number: 593-81-7
EINECS: 209-810-0
Merck Index: 12,09841

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
Specific optical rotation +26.5° to +30.5° (20°C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR