Fluo-3, pentaammonium salt calcium indicator

Supplier: AAT BIOQUEST INC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-[2-Amino-5-(2,7-dichloro-6-hydroxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid .5NH3Fluo-3

AATB21018EA 195 GBP
AATB21018
Fluo-3, pentaammonium salt calcium indicator
Fluo-3, pentaammonium salt

Calcium measurement is critical for numerous biological investigations.


  • Intracellular probe
  • Calcium-dependent fluorescence

MW: 854.69 g/mol
CAS Number: 339221-91-9
ADR: 5,II
https://uk.vwr.com/stibo/web/std.lang.all/31/65/43933165.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR