Linezolid, white

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Zyvox(S)-N-((3-(3-fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl)methyl)acetamide

PROOTRCL466500-50EA 0 GBP
PROOTRCL466500-50
Linezolid, white
Linezolid
Formula: C₁₆H₂₀FN₃O₄
MW: 337.4 g/mol
Melting Pt: 176...180 °C
MDL Number: MFCD00937825
CAS Number: 165800-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR