Biotin hydrazide ≥95%

Supplier: AAT BIOQUEST INC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide

AATB3007EA 102 GBP
AATB3007
Biotin hydrazide ≥95%
Biotin hydrazide

Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins.

Formula: C₁₀H₁₈N₄O₂S
MW: 258.34 g/mol
MDL Number: MFCD00078532
CAS Number: 66640-86-6
ADR: 5,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR