BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) ≥95%
Supplier: AAT BIOQUEST INC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
BCEF acid
AATB21201EA
129
GBP
AATB21201
BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) ≥95%
BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein)
Intracellular pH plays an important modulating role in many cellular events, including cell growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, cell adhesion and other cellular processes.
Formula:
C₂₇H₂₀O₁₁ MW: 520.44 g/mol |
MDL Number:
MFCD00036970 CAS Number: 85138-49-4 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...