Anti-CA125 Mouse Monoclonal Antibody [clone: X75]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNB600-1468EA 0 GBP
NOVUNB600-1468
Anti-CA125 Mouse Monoclonal Antibody [clone: X75]
Antibodies
MUC16 (CA125) is a serum marker that is used routinely in gynecologic practice to monitor patients with ovarian cancer.

CA125 is a mullerian duct differentiation antigen that is over expressed in epithelial ovarian cancer cells and secreted into the blood, although its expression is not entirely confined to ovarian cancer. Most biochemical studies have concluded that MUC16 is a high molecular mass glycoprotein, although estimates of its size range from 200 to 2000 kDa with smaller 'subunits' being described by some investigators.

Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Type: Primary
Antigen: CA125
Clonality: Monoclonal
Clone: X75
Conjugation: Unconjugated
Epitope: B1 epitope group
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR