Claritas PPT® Multi-element Standards Solution 4 for ICP and ICP-MS Instrumentation
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Claritas PPT®
COLE04568-86EA 0 GBP
COLE04568-86
Claritas PPT® Multi-element Standards Solution 4 for ICP and ICP-MS Instrumentation
Standards Cationic Standards Multi-Element Standards
Calibrate ICP-MS instrumentation prior to analysis.
  • Designed to contain virtually every element in the mass spectrum for concentration verification checks
Certifications: Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance.