Claritas PPT® Multi-element Standards Solution 2 for ICP and ICP-MS Instrumentation
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Claritas PPT®
COLE04568-82EA 0 GBP
COLE04568-82
Claritas PPT® Multi-element Standards Solution 2 for ICP and ICP-MS Instrumentation
Standards Cationic Standards Multi-Element Standards
Tune and calibrate ICP-MS instrumentation prior to analysis.
Multi element standards for Aluminum, Arsenic, Barium, Beryllium, Bismuth, Cadmium, Calcium, Cesium, Chromium, Cobalt, Copper, Gallium, Indium, Iron, Lead, Lithium, Magnesium, Manganese, Nickel, Potassium, Rubidium, Selenium, Silver, Sodium, Strontium, Thallium, Uranium, Vanadium, Zinc.
Certifications: Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance.