ABL90 FLEX PLUS Blood Gas Analyser
Supplier: Radiometer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RADCABL90FLEXPLUSEA 25130 GBP
RADCABL90FLEXPLUS
ABL90 FLEX PLUS Blood Gas Analyser
Clinical Diagnostic Supplies Clinical Analysers
ABL90 FLEX PLUS is the blood gas analyser of choice when the workflow demands quick analysis, efficiency and extensive insights from small sample sizes.
  • Fast analysis
  • Reliable results
  • Easy to use and maintain
Fast analysis of up to 19 parameters:
The ABL90 FLEX PLUS delivers test results in just 35 seconds on up to 19 vital parameters, including creatinine and urea - all from one blood sample of only 65 µl*. With a high uptime of more than 23,5 hours per day and only 60 seconds between each sample measurement, the analyser is quickly ready for you to test the next sample*.
Reliable results:
The ABL90 FLEX PLUS delivers lab-quality results at the point of care for more confident treatment decisions. Continuous automatic quality management (AQM) provides automatic quality control, continuous checks and automatically initiates and documents corrective actions. In addition, automatic patient registration, automatic sample mixing, and safePICO syringes help reduce the risk of patient sample mix up and preanalytical errors.
Easy to use and maintain:
The ABL90 FLEX PLUS blood gas analyser is designed to be easy to use and maintain so your staff spends less time in front of the analyser. Instructional videos provide on-screen guidance for operation and maintenance.
And, with only two consumables to replace, remote service and full IT connectivity, your ABL90 FLEX PLUS blood gas analyser is ready when you are - whether it’s in the lab or at the point of care.
Built-in cybersecurity:
The ABL90 FLEX PLUS is built on the Radiometer cybersecurity model, which works alongside hospital security measures in a series of defense layers. These defense layers help prevent cyberattacks from compromising patient data and analyser performance. For extra protection, Security packages update analysers’ security levels throughout their lifetime.