COULTER DxH Cleaning Agent
Supplier: Beckman Coulter
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BECM628023EA 0 GBP
BECM628022 BECM628023
COULTER DxH Cleaning Agent
Cleaning Agents Detergents and Surface Cleansers
A cleaning agent for use on UniCel DxH Coulter cellular analysis systems for components that come in contact with blood samples.
  • Ready to use
  • Store at 2 to 25 °C
This azide-free, formaldehyde-free cleaner contains a proteolytic enzyme that aids in the removal of protein buildup. The cleaning reagent dissolves proteins at apertures as well as at aspiration and dispensing components of the systems.
Caution: For in vitro diagnostic use.