Environmental Express Digestion Tube
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Environmental Express
COLEUC474EA 0 GBP
COLEUC474
Environmental Express Digestion Tube
Tubes Digestion Tubes
Disposable digestion tubes eliminate transfer steps.
  • Tubes that act as a digestion vessel or storage container and fit perfectly into the HotBlock and AutoBlock systems
  • Low metals background and EPA approved for digestions
  • Accessories include watch glasses, reflux caps, and transfer/storage racks
Approved for hot plate digestions and certified for low metals background.