Environmental Express FlipMate Filtration Assemblies
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Environmental Express FlipMate
COLESC0601EA 0 GBP
COLESC0601
Environmental Express FlipMate Filtration Assemblies
Filters Membrane Filter Units
Assemblies connect directly to your existing digestion cups.
  • Sample is ready in just seconds
  • Convenient, economical method for filtration
  • Produce quality results with clean samples
The FlipMate assemblies easily filter your sample with the use of two threaded digestion cups, a little tubing, and vacuum pressure. The sample is placed in one cup, the filter assembly threaded onto the top of the cup, then the cup and filter are flipped, the receiving cup attached, vaccum tubing attached, vaccum pressure applied, and the sample is then pulled from the sample cup, through the filter assembly, and into the empty cup. Digestions cups, tubing, and vacuum source are not included, filtration assembly only.