2-Hydroxy-4-methoxy-5-sulphobenzophenone ≥97.0%
Supplier: Spectrum ChemicalTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
Benzophenone-4
, 5-Benzoyl-4-hydroxy-2-methoxybenzenesulphonic acid
, 2-Hydroxy-4-methoxybenzophenone-5-sulphonic Acid
Benzophenone-4 is also known as sulisobenzone and is typically an ingredient in most sunscreens. It is used as a skin protectant from damage by short-wave UVA ultraviolet light and UVB.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₄H₁₂O₆S MW: 308.30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00024962 CAS Number: 4065-45-6 |
Specification Test Results
Assay (Dry Basis) | 97.0 - 103.0% |
Extinction Coefficient in 95% Ethanol at 285 NM | 460 min. |
pH of a 1% Solution | 1.2 - 2.2 |
Color (Gardner; 10% W/V in Water) | max. 4.0 |
Solubility of 10% W/V in Water | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...