Petrolatum, yellow USP
Supplier: Spectrum Chemical
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SPCMPE130-500GMEA
146
GBP
SPCMPE130-500GM
Petrolatum, yellow USP
Petrolatum
Petrolatum, Yellow, USP is a carrier or vehicle excipient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage Temperature:
Ambient |
MDL Number:
MFCD00147839 CAS Number: 8009-03-8 |
Specification Test Results
Stabilizer (Butylhydroxytoluene) | max. 0.002% |
Specific Gravity @60 C | 0.815-0.880 |
Melting Range | 38 - 60 °C |
Color | To Pass Test |
Acidity | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...