Nɑ-Acetyl-L-glutamine
Supplier: Spectrum Chemical
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ac-Gln-OH
, N-Acetyl-L-glutamine
, Ac-L-Gln-OH
, N-Ac-Gln-OH
SPCMA2393-100KGBLEA
0
GBP
SPCMA2393-100KGBL
Nɑ-Acetyl-L-glutamine
Nalpha-Acetyl-L-glutamine
N-Acetyl-L-glutamine is used as a skin conditioning agent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₇H₁₂N₂O₄ MW: 188.18 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00038159 CAS Number: 2490-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...