Propyl acetate ≥98.0% (by GC)

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Acetic acid propyl ester

FLUOA0044.500MLEA 0 GBP
FLUOA0044.500ML
Propyl acetate ≥98.0% (by GC)
Propyl acetate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 101°C
Flash Pt: 13°C
MDL Number: MFCD00009372
CAS Number: 109-60-4
EINECS: 203-686-1
UN: 1276
ADR: 3,II
https://uk.vwr.com/stibo/web/std.lang.all/74/89/28927489.jpg

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR