1,5-Diaminopentane ≥98.0% (by titrimetric analysis)

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Cadaverine1,5-PentanediamineCadavarinePentamethylendiamine

FLUOD0108.5MLEA 0 GBP
FLUOD0108.5ML
1,5-Diaminopentane ≥98.0% (by titrimetric analysis)
1,5-Diaminopentane
Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 180°C
Flash Pt: 63°C
MDL Number: MFCD00008239
CAS Number: 462-94-2
EINECS: 207-329-0
UN: 2735
ADR: 8,II
https://uk.vwr.com/stibo/web/std.lang.all/44/79/28974479.jpg

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR