2-Phenyl-1,3-propanediol ≥98.0% (by GC)

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

FLUOP1249EA 0 GBP
FLUOP1249
2-Phenyl-1,3-propanediol ≥98.0% (by GC)
2-Phenyl-1,3-propanediol
Formula: HOH₂CCH(C₆H₅)CH₂OH
MW: 152.19 g/mol
MDL Number: MFCD00236056
CAS Number: 1570-95-2
EINECS: 411-810-2
https://uk.vwr.com/stibo/web/std.lang.all/18/82/28991882.jpg

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR