2-Phenyl-2-oxazoline ≥98.0% (by GC, titration analysis)

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Phenyl(2-oxazoline)(4,5-Dihydro-2-oxazolyl)benzene4,5-Dihydro-2-phenyloxazole

FLUOD2155EA 0 GBP
FLUOD2155
2-Phenyl-2-oxazoline ≥98.0% (by GC, titration analysis)
2-Phenyl-2-oxazoline
Formula: C₉H₉NO
MW: 147.1739 g/mol
Boiling Pt: 243°C
Storage Temperature: Refrigerator
MDL Number: MFCD00075541
CAS Number: 7127-19-7
https://uk.vwr.com/stibo/web/std.lang.all/89/47/28978947.jpg

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR