Potassium carbonate sesquihydrate ≥99% ACS

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

SIAL243558-1KGEA 140 GBP
SIAL243558-1KG
Potassium carbonate sesquihydrate ≥99% ACS
Potassium carbonate sesquihydrate
Formula: CK₂O₃·1,5H₂O
MW: 165.23 g/mol
Melting Pt: 891 °C
Density: 2.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149925
CAS Number: 6381-79-9
EINECS: 209-529-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR