Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate), Pierce™
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Sulpho succinimidyl 4-(ρ-maleimidophenyl) butyrate
, Sulfo SMPB
, Sulphosuccinimidyl 4-(N-maleimidophenyl)butyrate
Pierce™
PIER22317EA
305
GBP
PIER22317
Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate), Pierce™
Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Thermo Scientific Pierce Sulfo-SMPB is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length aromatic spacer arm (11.6 angstroms).
- Reactive groups: sulfo-NHS ester and maleimide
- Reactive towards: amino and sulfhydryl groups
- Extended chain length limits steric hindrance
- Non-cleavable
- Water-soluble (compare to SMPB)
- Membrane impermeable, allowing for cell surface labeling
Caution: For Research Use Only. Not for use in diagnostic procedures.
Formula:
C₁₈H₁₅N₂O₉S·Na MW: 458.374 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00054981 CAS Number: 92921-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...