Bromine solution R Reag. Ph. Eur. 1012401

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

85715.180EA 97.4 GBP
85715.180
Bromine solution R Reag. Ph. Eur. 1012401
Bromine

The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for Purified Water. Short shelf life of max. 3 months


Starting materials: Bromine R / Potassium bromide R / Water R

Formula: Br₂
MW: 159.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010896
CAS Number: 7726-95-6
UN: 1744
ADR: 8(6.1),I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR