Water, STERI-WATER® USP, purified, sterile

Supplier: VELTEK ASSOCIATES INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

VELTSTWA-5GEA 0 GBP
VELTSTWA-5G
Water, STERI-WATER® USP, purified, sterile
Water

Sterile water for chemical formulation, cleaning, and rinsing.


Filtered at 0,2 µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.


Caution: Not for human or animal injection, diagnostic, or therapeutic use.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR