Soybean Trypsin inhibitor
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIMI1441-1G
BIMI1441-250MG
BIMI1441-5G
APOSBIMI1441-1GEA
153
GBP
APOSBIMI1441-1G
APOSBIMI1441-250MG
APOSBIMI1441-5G
Soybean Trypsin inhibitor
Proteins and Peptides
Assay >10000 U/mg lyophilizedInhibitor of Trypsin, Chymotrypsin, Plasmin and otherproteolytic enzymes.