2,2,2-Trifluoroethyl methyl ether 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC7375-10G PC7375-25G
APOSPC7375-10GEA 158 GBP
APOSPC7375-10G APOSPC7375-25G
2,2,2-Trifluoroethyl methyl ether 97%
2,2,2-Trifluoroethyl methyl ether
Formula: C₃H₅F₃O
MW: 114.07 g/mol
Boiling Pt: 30 °C
Density: 1.1661 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00236114
CAS Number: 460-43-5
UN: 1993
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR