2-Chloro-3-(trifluoromethyl)phenylboronic acid 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
APOSPC7131-1GEA
47.4
GBP
APOSPC7131-1G
2-Chloro-3-(trifluoromethyl)phenylboronic acid 98%
2-Chloro-3-(trifluoromethyl)phenylboronic acid
Formula:
C₇H₅BClF₃O₂ MW: 224.3726 g/mol Melting Pt: 77…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258745 CAS Number: 957061-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...