Difluoromethyl-2,2,3,3-tetrafluoropropyl ether 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC6868B-1G PC6868B-5G
APOSPC6868B-1GEA 99.9 GBP
APOSPC6868B-1G APOSPC6868B-5G
Difluoromethyl-2,2,3,3-tetrafluoropropyl ether 97%
Difluoromethyl-2,2,3,3-tetrafluoropropyl ether
Formula: C₄H₄F₆O
MW: 182.07 g/mol
Boiling Pt: 69 °C
Density: 1.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236119
CAS Number: 35042-99-0
UN: 3271
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR