Perfluorooctadecanoic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

APOSPC6136-250MGEA 47.4 GBP
APOSPC6136-250MG APOSPC6136-25G APOSPC6136-5G APOSPC6136-1G
Perfluorooctadecanoic acid
Perfluorooctadecanoic acid
Formula: C₁₈HF₃₅O₂
Boiling Pt: 235 °C/100mmHg
Melting Pt: 163…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00198021
CAS Number: 16517-11-6
EINECS: 240-582-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR