4-Fluoro-2-(trifluoromethoxy)aniline 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-fluoro-2-(trifluoromethoxy)aniline

PC57037-1G PC57037-5G
APOSPC57037-1GEA 206 GBP
APOSPC57037-1G APOSPC57037-5G
4-Fluoro-2-(trifluoromethoxy)aniline 98%
4-Fluoro-2-(trifluoromethoxy)aniline
Formula: C₇H₅F₄NO
MW: 195.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527447
CAS Number: 123572-66-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR