5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-(Trifluoromethyl)-7-azaindole-2-carboxylic acid

PC57033-1G PC57033-250MG
APOSPC57033-1GEA 416 GBP
APOSPC57033-1G APOSPC57033-250MG
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid 95%
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid
Formula: C₉H₅F₃N₂O₂
MW: 230.15 g/mol
Melting Pt: 328…330 °C
Storage Temperature: Ambient
MDL Number: MFCD15529028
CAS Number: 784144-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR