1,1,1,3,3-Pentafluoropropane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC5702D-100G PC5702D-25G PC5702D-500G
APOSPC5702D-100GEA 746 GBP
APOSPC5702D-100G APOSPC5702D-25G APOSPC5702D-500G
1,1,1,3,3-Pentafluoropropane 98%
1,1,1,3,3-Pentafluoropropane
Formula: C₃H₃F₅
MW: 134.05 g/mol
Boiling Pt: 15 °C
Melting Pt: –102.1 °C
Density: 1.32 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 460-73-1
UN: 3163
ADR: 2.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR