2-Fluoro-5-nitropyridine 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
APOSPC48306-25GEA
70.4
GBP
APOSPC48306-25G
2-Fluoro-5-nitropyridine 97%
2-Fluoro-5-nitropyridine
Formula:
C₅H₃FN₂O₂ Boiling Pt: 86…87 °C/7mmHg Melting Pt: 17…21 °C Density: 1.41 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03095059 CAS Number: 456-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...