6-(2,2,2-Trifluoroethoxy)nicotinic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC32063-10G PC32063-1G PC32063-5G
APOSPC32063-10GEA 196 GBP
APOSPC32063-10G APOSPC32063-1G APOSPC32063-5G
6-(2,2,2-Trifluoroethoxy)nicotinic acid
6-(2,2,2-Trifluoroethoxy)nicotinic acid
Formula: C₈H₆F₃NO₃
Melting Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00110708
CAS Number: 175204-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR