1-Fluorocyclohexanecarboxylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC2955-250MG
APOSPC2955-250MGEA 162 GBP
APOSPC2955-250MG
1-Fluorocyclohexanecarboxylic acid
1-Fluorocyclohexanecarboxylic acid
Formula: C₇H₁₁FO₂
MW: 146.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08751334
CAS Number: 117169-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR