3-Nitro-4-(trifluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PC1558-100G
APOSPC1558-100GEA
147
GBP
APOSPC1558-100G
3-Nitro-4-(trifluoromethoxy)aniline 98%
3-Nitro-4-(trifluoromethoxy)aniline
Formula:
C₇H₅F₃N₂O₃ Melting Pt: 90…92 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840207 CAS Number: 2822-50-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...