2-Bromo-6-fluorobenzonitrile 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC10431-100G PC10431-25G PC10431-5G
APOSPC10431-100GEA 115 GBP
APOSPC10431-100G APOSPC10431-25G APOSPC10431-5G
2-Bromo-6-fluorobenzonitrile 98%
2-Bromo-6-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD06797753
CAS Number: 79544-27-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR