2-Bromo-6-fluorobenzamide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC10430-25G PC10430-5G
APOSPC10430-25GEA 157 GBP
APOSPC10430-25G APOSPC10430-5G
2-Bromo-6-fluorobenzamide
2-Bromo-6-fluorobenzamide
Formula: C₇H₅BrFNO
MW: 218.0231 g/mol
Melting Pt: 154…156 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07368694
CAS Number: 107485-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR