2-Iodo-4-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Iodo-α,α,α-trifluoro-p-toluidine
, 4-Amino-3-iodobenzotrifluoride
, 4-Amino-3-iodo-α,α,α-trifluorotoluene
APOSPC1039-5GEA
10.6
GBP
APOSPC1039-5G
2-Iodo-4-(trifluoromethyl)aniline 98%
2-Iodo-4-(trifluoromethyl)aniline
Formula:
C₇H₅F₃IN MW: 287.02 g/mol Melting Pt: 51…53 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02683097 CAS Number: 163444-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...