2,3,3,3-Tetrafluoropropene-1 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

PC0987-100G PC0987-25G PC0987-5G
APOSPC0987-100GEA 474 GBP
APOSPC0987-100G APOSPC0987-25G APOSPC0987-5G
2,3,3,3-Tetrafluoropropene-1 97%
2,3,3,3-Tetrafluoropropene-1
Formula: C₃H₂F₄
MW: 114.04 g/mol
Boiling Pt: –28 °C
Melting Pt: –152.24 °C
Storage Temperature: Ambient
MDL Number: MFCD01320735
CAS Number: 754-12-1
UN: 3161
ADR: 2.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR