2H,3H-Perfluoropentane 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PC0375-100G PC0375-1KG PC0375-250G
APOSPC0375-100GEA 63.1 GBP
APOSPC0375-100G APOSPC0375-1KG APOSPC0375-250G
2H,3H-Perfluoropentane 98%
2H,3H-Perfluoropentane

Non-Ozone depleting replacement for FC-113.

Formula: C₅H₂F₁₀
Boiling Pt: 55 °C
Melting Pt: –80 °C
Density: 1.6 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798129
CAS Number: 138495-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR