1-Chlorobenzotriazole 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR960666-1G OR960666-25G OR960666-5G
APOSOR960666-1GEA 78.8 GBP
APOSOR960666-1G APOSOR960666-25G APOSOR960666-5G
1-Chlorobenzotriazole 97%
1-Chlorobenzotriazole
Formula: C₆H₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD00054541
CAS Number: 21050-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR