4,5,6,7-Tetrahydro-1,2-benzisoxazol-3-ylmethanol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR943742-1G OR943742-250MG
APOSOR943742-1GEA 469 GBP
APOSOR943742-1G APOSOR943742-250MG
4,5,6,7-Tetrahydro-1,2-benzisoxazol-3-ylmethanol 95%
4,5,6,7-Tetrahydro-1,2-benzisoxazol-3-ylmethanol
Formula: C₈H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD08701254
CAS Number: 893638-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR