(R)-1-Phenyl-1,3-propanediol 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR925648-1G
OR925648-250MG
APOSOR925648-1GEA
521
GBP
APOSOR925648-1G
APOSOR925648-250MG
(R)-1-Phenyl-1,3-propanediol 95%
(R)-1-Phenyl-1,3-propanediol
Formula:
C₉H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00145213 CAS Number: 103548-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...